PCR and qPCR
Shop by PCR and qPCR Methods
Preparing samples for PCR or qPCR typically requires extraction and purification of nuclear material. Depending on the sample source, this may involve mechanical manipulation via homogenization or require the use of resins or beads to help bind the nucleic acids prior to purification. Prepare your samples with the help of DNA and RNA extraction, isolation, and purification kits, plus reagents, buffers, and more.
Top Categories
Amplification is performed by combining a DNA segment, polymerase, nucleotide solution, and primers in a PCR tube or microplate. A thermal cycler is used to perform the following steps, repeating the process 35 to 40 times.
Denaturation: Hydrogen bonds are broken, DNA separates.
Annealing: Primers and polymerase bind to the DNA.
Extension: Nucleotides pair with the DNA to form a new complementary strand.
Top Categories
In real-time PCR, RNA is converted to cDNA and then amplified using PCR. Common real-time PCR applications detect expressed genes, examine transcript variants, and create cDNA templates for cloning and sequencing procedures. Find polymerases, oligonucleotides, master mixes, and more below to support your real-time PCR assays.
Top Categories
Real-time PCR is used to detect specific DNA sequences. This method uses fluorescent dyes or probes to determine the amount of amplified material in your sample. Find a variety of master mixes, plus real-time PCR instruments, assays, and kits for quantifying your qPCR results.
Top Categories
Find HEPA-filtered PCR enclosures to help keep contaminating particulates out of your workspace. Shop single-use PCR tubes and tube strips, nucleic acid microplates, PCR enclosures, and other labware that is certified free of DNAse, RNAse, DNA, and other potential contaminants.
Top Categories
Related Resources
Other Applications
Molecular Biology Methods
Explore solutions for isolation and purification, amplification, transfection, and more.
Protein Biology Methods
Browse isolation, purification, separation, and detection essentials.