Analytical Testing Methods
High-Quality Analytical Testing Products from Top Brands
Analytical testing methods help Iabs and researchers identify and monitor acidity and ion concentrations, water content, and many other properties of a range of sample types.
Analytical techniques such as Karl Fischer titration, as well as pH and ASTM methods, are frequently used in water testing, environmental analysis, and food and beverage testing. They are also used in pharmaceutical production and the production of cosmetics, petroleum, building materials, and more, as well as in clinical research.
Accurate, consistent outcomes are mandatory and efficiency is just as important. The analytical testing chemicals and lab products you choose help to deliver the outcomes you need and boost confidence that your samples won't be wasted.
1. pH and Electrochemistry
pH and electrochemistry meters and related products are used to assess acidity and alkalinity, conductivity, specific ion concentrations, and dissolved oxygen in water, wastewater, and other solutions.
Electrodes must be carefully stored and maintained for optimal performance. As such it is important to select the right storage bottles and electrode filling, cleaning, rinsing, and storage accessories.
Electrochemistry meters also require regular calibration and maintenance. Standard and certified pH buffers are available, as well as conductivity and specific ion standards. Other accessories include electrode supports, bulb guards, cables, connectors, and adapters.
Top Categories
Resources
Fisher Scientific Certified pH Buffers
- Filename
- 17312_Certified_pH_Buffers_spex.pdf
- Size
- 268 KB
- Format
- application/pdf
Thermo Scientific and Fisher Chemical Electrochemistry Analytical Testing Products
- Filename
- 17312_Electrochemistry_Workflow.pdf
- Size
- 894 KB
- Format
- application/pdf
2. Karl Fischer Titration
Karl Fischer titration is used with a range of sample types to determine trace amounts of water in samples. Labs and researchers use two types of Karl Fischer titration based on a sample's properties.
Coulometric KF titration is ideal for liquid and gaseous samples with low water content (up to 2%). Water content is measured by electrical current and the instrument's cell is made from glass with ground joint connections that ensure a tight seal against atmospheric moisture.
Volumetric KF titration is used with solid, liquid, and gaseous samples with high water content (over 2%). Water content is measured by burette instead of electrical current and the instrument's cell construction differs from a coulometric titrator.
Top Categories
Resources
Fisher Chemical Aqualine Reagents Brochure
- Filename
- 17312_Fisher_Chemical_Aqualine_Reagents.pdf
- Size
- 1 MB
- Format
- application/pdf
Honeywell Analytical Testing Products Data Sheet
- Filename
- 17312_Product_Overview_Guide_Hydranal.pdf
- Size
- 1 MB
- Format
- application/pdf
Honeywell Water Determination Hydranal Brochure
- Filename
- 17312_Water_Determination_Hydranal.pdf
- Size
- 949 KB
- Format
- application/pdf
Honeywell Free of Imidazole Hydranal Reagents
- Filename
- 17312_Hydranal_Free_Of_Imidazole_2022.pdf
- Size
- 2 MB
- Format
- application/pdf
3. Trace Elemental Analysis
In chemistry, elemental analysis is the activity of determining the chemical elements of a compound by using chemical analysis, organic analysis, spectroscopy or atomic absorption spectrometry, among other methods. Sometimes, the sample's isotopic composition is also defined.
The quantity determination of a specific element within a sample material (soil, waste, water, minerals, chemical compounds etc) can be performed on a solid, liquid or gas and can be divided into two categories:
- Qualitative analysis: determines which elements are present
- Quantitative analysis: determines how much of each element is present
We can offer a complete line of different grade reagents and elemental standards to meet the most challenging applications.
Different grades of products are available depending on the sensitivity you need and the technology you use, e.g.: AAS, GFAAS, ICP, ICP-OES, and ICP-MS.
Top Categories
Resources
Fisher Chemical High-Purity Acids
- Filename
- 17914_Fisher_Chemical_High_Purity_Acids.pdf
- Size
- 1 MB
- Format
- application/pdf
Thermo Scientific Specpure Analytical Standards
- Filename
- 17914_Thermo_Scientific_Specpure_Analytical_Standards.pdf
- Size
- 1 MB
- Format
- application/pdf
SPEX Speciation Standards
- Filename
- 17914_SPEX_Speciation_Standards.pdf
- Size
- 412 KB
- Format
- application/pdf
SPEX ICH/Global Compliance
- Filename
- 17914_SPEX_ICH_Global_Compliance.pdf
- Size
- 431 KB
- Format
- application/pdf
SPEX 1 PPM Single Element Standards
- Filename
- 17914_SPEX_1_PPM_Single_Element_Standards.pdf
- Size
- 406 KB
- Format
- application/pdf
SPEX Heavy Metals and Minerals Testing Kits
- Filename
- 17914_SPEX_Heavy_Metals_and_Minerals_Testing_Kits.pdf
- Size
- 517 KB
- Format
- application/pdf
SPEX SPEX 30 mL Single-Element Standards
- Filename
- 17914_SPEX_30_mL_Single_Element_Standards.pdf
- Size
- 391 KB
- Format
- application/pdf
SPEX USP <232>, <233> and <2232> Elemental Impurities
- Filename
- 17914_SPEX_Elemental_Impurities.pdf
- Size
- 650 KB
- Format
- application/pdf
4. ASTM Methods
American Society for Testing and Materials (ASTM) test methods are definitive procedures that produce a result. They usually include a detailed description of a procedure for determining a property or constituent of a material, an assembly of materials, or a product. These details include the apparatus, test specimen, procedure, and calculations needed for satisfactory results.
ASTM test methods represent a consensus about the best currently available procedure and are supported by experience and data from cooperative tests.
The ASTM publishes six types of standards: test method, specification, classification, practice, guide, and terminology.
Resources
The Four Ws of ASTM
- Filename
- 17312_The_Four_Ws_of_ASTM.pdf
- Size
- 285 KB
- Format
- application/pdf
Solvents for Trace Analysis by UHPLC-MS White Paper
- Filename
- 17312_Solvents_for_Trace_Analysis_by_UHPLC-MS_White_Paper.pdf
- Size
- 551 KB
- Format
- application/pdf